Learn More
Cobalt(II,III) oxide, nanopowder, 99% (metals basis)
CAS: 1308-06-1 | Co3O4 | 240.80 g/mol
Supplier: Thermo Scientific Chemicals 04466122
Description
Cobalt(II,III) oxide is used in enamels, pigments, porcelain colorants, semiconductors, ceramics, catalysis, grinding wheels and other fields as an important inorganic material. It is a useful oxidant in chemical industry and as catalyst for organic synthesis. It is widely used as catalyst carrier and as electrode active material. It is also used in senior goggles, other filter materials, in temperature and gas sensors, in electronic ceramics, in lithium ion battery electrode materials, magnetic materials, electrochromic devices and solar energy absorbers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Environmental benefits include:
- Recyclable
Specifications
Cobalt (II,III) oxide | |
1308-06-1 | |
Powder | |
(metals basis) | |
MFCD00010939 | |
14,2429 | |
Soluble in acids and alkali. Insoluble in water. | |
[O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3] | |
240.80 | |
240.8 | |
6.11 g/mL |
900°C (decomposition) | |
Black | |
100 g | |
Co3O4 | |
UN3077 | |
oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt | |
UBEWDCMIDFGDOO-UHFFFAOYSA-N | |
oxocobalt;oxo(oxocobaltiooxy)cobalt | |
11651651 | |
99% |
Chemical Identifiers
1308-06-1 | |
240.80 | |
UBEWDCMIDFGDOO-UHFFFAOYSA-N | |
11651651 | |
[O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3] |
Co3O4 | |
MFCD00010939 | |
oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobaltShow More | |
oxocobalt;oxo(oxocobaltiooxy)cobalt |
Safety and Handling
GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
H317-H334-H350i
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 215-157-2
RTECSNumber : GG2500000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only