Learn More
Cobalt(II) acetate tetrahydrate, ACS, 98.0-102.0%
CAS: 6147-53-1 | C4H14CoO8 | 249.081 g/mol
Supplier: Thermo Scientific Chemicals 04434422
Description
Cobalt(II) acetate tetrahydrate is used as an industrial catalyst to harden paints and varnishes, an active catalyst for oxidation and esterification reactions. It is also used to prepare metal complexes having unusual coordination geometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cobalt (II) acetate tetrahydrate | |
6147-53-1 | |
Crystalline | |
C4H14CoO8 | |
MFCD00149569 | |
3731397 | |
cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441 | |
ZBYYWKJVSFHYJL-UHFFFAOYSA-L | |
cobalt(2+);diacetate;tetrahydrate | |
61372 | |
98.0 to 102.0% | |
Odorless | |
Hygroscopic |
∼140°C (in -4H2O) | |
Red | |
100 g | |
Co(OOCCH3)2·4H2O | |
UN3077 | |
14,2433 | |
Soluble in water,alcohol,dilute acids and phenyl acetate. | |
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2] | |
249.081 | |
249.08 (177.02 Anhydrous) | |
ACS | |
1.542 | |
1.71 g/mL |
Chemical Identifiers
6147-53-1 | |
249.081 | |
ZBYYWKJVSFHYJL-UHFFFAOYSA-L | |
61372 | |
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2] |
C4H14CoO8 | |
MFCD00149569 | |
cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441Show More | |
cobalt(2+);diacetate;tetrahydrate |
Safety and Handling
GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.
P201-P202-P261-P271-P272-P280g-P281-P285-P302+P352-P304+P340-P308+P313-P333+P313-P342+P311-P363-P501c
H302-H317-H334-H335-H341-H350i-H360F
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 200-755-8
RTECSNumber : AG3325000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only