missing translation for 'onlineSavingsMsg'
Learn More

Brady™ CleanLift™ Thermal Transfer Printable Labels - 1 in. Core

One-inch core materials to fit inside label printers. Brady CleanLift Thermal Transfer Printable Labels - 1 in. Core are specific to laboratory, healthcare and general industrial markets as well as aerospace, defence and mass transit markets.

Supplier:  Brady™ THT9549825SC

 View more versions of this product

Catalog No. 19-109-430


Only null left
Explore available promotions

Description

Description

  • For use with thermal transfer printers
  • Made of B-498 vinyl cloth with repositionable rubber adhesive
Greener Choice Claims
  • Environmental benefits include:
    • Source Reduction
Specifications

Specifications

UL Recognized
Rubber, Repositionable
Semi-gloss
Thermal Transfer Printable Labels - 1 in. Core
BBP11 Label Printer, Brady 1244 and 1344 Series
0.800 x 1.437 in. (W x H)
0.0073 in.
Suggested ribbon series: R4300; R4800; R6000; R6200
White
Vinyl Cloth (B-498)
Barcode labeling, Cable and wire marking, Circuit Board ID - Through hole-top, Component and equipment ID, General and industrial ID
Rectangle
20.32 x 36.49 mm (W x H)
20.32 x 36.49 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions