missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Hexene, 96%

CAS: 7688-21-3 | C6H12 | 84.16 g/mol

Supplier:  Thermo Scientific Chemicals H5350606

 View more versions of this product

Catalog No. AAH5350606


Only null left
Explore available promotions

Description

Description

cis-2-Hexene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-2-Hexene is used as an organic chemical synthesis intermediate.

Solubility
Immiscible in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 231-697-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only