missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady Chemical Resistant Polypropylene Labels for BradyJet™ J7300 Color Label Printer

Create full-color identification and barcode labels that outlast liquid nitrogen, autoclave conditions and harsh chemicals.
Supplier: Brady J1367425J
Description
Create full-color identification and barcode labels that outlast liquid nitrogen, autoclave conditions and harsh chemicals. InkJet-receptive label material is ideal for full-color laboratory identification on vials, centrifuge tubes and test tubes.
- Labels withstand extreme temperatures from -112° to 158°F (-80° to 70°C).
- Permanent acrylic adhesive bonds tightly to smooth surfaces like glass, copper and stainless steel. RoHS compliant to 2011/65/EU and 2015/863.
- Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup and seamless software interaction.
- For use with the BradyJet™ J7300 Color Label Printer.
- Ink cartridges are required to print on this material, use part numbers P-CMY-1 and P-K-1.
Greener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
ASTM D 1000;ASTM D 2979 | |
None | |
Matte | |
Polypropylene | |
Ink Jet | |
Non-sterile | |
J7300 | |
1 | |
1000 Labels | |
-112°F to 158°F | |
0.005 in. | |
0.75 x 1 in. | |
19.05 mm | |
38.10 mm |
Permanent Acrylic | |
White | |
Die-cut | |
Single-side Printable | |
Low Temperature Labels | |
Barcode Labeling, Cold temperature Labeling, General Identification, Hazard Communication Identification, Laboratory Labeling | |
No Secondary Label | |
No Text | |
1000 Labels | |
-80°C to 70°C | |
0.127 mm | |
0.75 in. | |
1.5 in. |