missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cetilistat 98.0+%, TCI Americaâ„¢
$30.24 - $81.25
Chemical Identifiers
CAS | 282526-98-1 |
---|---|
Molecular Formula | C25H39NO3 |
Molecular Weight (g/mol) | 401.591 |
MDL Number | MFCD09839697 |
InChI Key | MVCQKIKWYUURMU-UHFFFAOYSA-N |
Synonym | 2-Hexadecyloxy-6-methyl-4H-3,1-benzoxazin-4-one |
PubChem CID | 9952916 |
IUPAC Name | 2-hexadecoxy-6-methyl-3,1-benzoxazin-4-one |
SMILES | CCCCCCCCCCCCCCCCOC1=NC2=C(C=C(C=C2)C)C(=O)O1 |
Chemical Identifiers
282526-98-1 | |
401.591 | |
MVCQKIKWYUURMU-UHFFFAOYSA-N | |
9952916 | |
CCCCCCCCCCCCCCCCOC1=NC2=C(C=C(C=C2)C)C(=O)O1 |
C25H39NO3 | |
MFCD09839697 | |
2-Hexadecyloxy-6-methyl-4H-3,1-benzoxazin-4-one | |
2-hexadecoxy-6-methyl-3,1-benzoxazin-4-one |
Specifications
282526-98-1 | |
White-Yellow | |
MFCD09839697 | |
2-Hexadecyloxy-6-methyl-4H-3,1-benzoxazin-4-one | |
CCCCCCCCCCCCCCCCOC1=NC2=C(C=C(C=C2)C)C(=O)O1 | |
401.591 | |
401.59 | |
Crystalline Powder |
74°C | |
C25H39NO3 | |
1 g | |
MVCQKIKWYUURMU-UHFFFAOYSA-N | |
2-hexadecoxy-6-methyl-3,1-benzoxazin-4-one | |
9952916 | |
≥98.0% (HPLC) | |
Cetilistat |
Safety and Handling
TSCA : No