missing translation for 'onlineSavingsMsg'
Learn More

Cedarwood Oil, Spectrum™ Chemical
GREENER_CHOICE

$250.50 - $813.43

Chemical Identifiers

CAS 68990-83-0
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18609590
View Documents
Spectrum Chemical Mfg Cor
CE108100ML
100 mL Amber Glass Bottle
Each for $250.50
Only null left
 
18609592
View Documents
Spectrum Chemical Mfg Cor
CE108500MLGL
500 mL Amber Glass Bottle
Each for $813.43
Only null left
 
Description

Description

Specifications

Chemical Identifiers

68990-83-0

Specifications

68990-83-0
Amber Glass Bottle
100 mL
1
1.510 -1.520
Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.