Learn More
Epredia™ Cassette II Biopsy Tissue Cassettes
For small tissue samples, these cassettes have a front-edge lid hinge with loose-packed lids that are available in 12 different color varieties
Supplier: Epredia™ B851738BL
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Small Squares | |
1000/Cs. | |
Separate Lid, Front Hinge | |
Blue |
Yes | |
Loose Packed (Lids) | |
No | |
Cassette II with Color-Matched Lids |