missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Carboxymethyl cellulose, sodium salt, average M.W. 250000 (DS=0.9)
CAS: 9004-32-4 | (C12 H14 O9 R6)n | 263.20 g/mol
$77.46 - $251.55
Chemical Identifiers
CAS | 9004-32-4 |
---|---|
Molecular Formula | (C12 H14 O9 R6)n |
Molecular Weight (g/mol) | 263.20 |
MDL Number | MFCD00081472 |
InChI Key | DPXJVFZANSGRMM-UHFFFAOYNA-N |
Synonym | carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn |
PubChem CID | 23706213 |
IUPAC Name | 2,3,4,5,6-pentahydroxyhexanal acetic acid sodium |
SMILES | [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC332621000
|
Thermo Scientific Chemicals
332621000 |
100 g | Glass bottle |
Each for $77.46
|
|
|||||
AC332620010
|
Thermo Scientific Chemicals
332620010 |
1 kg | Plastic bottle |
Each for $251.55
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9004-32-4 | |
263.20 | |
DPXJVFZANSGRMM-UHFFFAOYNA-N | |
23706213 | |
[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O |
(C12 H14 O9 R6)n | |
MFCD00081472 | |
carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tnShow More | |
2,3,4,5,6-pentahydroxyhexanal acetic acid sodium |
Specifications
White to Beige | |
Crystalline Powder | |
(C12 H14 O9 R6)n | |
15, 1830 | |
100 g | |
[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O | |
263.20 | |
≥99.5% | |
10% max. (as packed) (3 to 5 g, 105°C, 2 hrs) | |
2300mPa·s, 2% sol. |
6.5 to 8 (1% soln.) | |
9004-32-4 | |
MFCD00081472 | |
carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn | |
DPXJVFZANSGRMM-UHFFFAOYNA-N | |
2,3,4,5,6-pentahydroxyhexanal acetic acid sodium | |
23706213 | |
Authentic | |
Glass bottle | |
Carboxymethylecellulose, sodium salt, Average M.W. 250000 (DS=0.9) |
RUO – Research Use Only