missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Carboxymethyl cellulose, sodium salt, average M.W. 250000 (DS=0.7)

CAS: 9004-32-4 | (C12 H14 O9 R6)n | 263.20 g/mol

$74.28 - $218.43

Chemical Identifiers

CAS 9004-32-4
Molecular Formula (C12 H14 O9 R6)n
Molecular Weight (g/mol) 263.20
MDL Number MFCD00081472
InChI Key DPXJVFZANSGRMM-UHFFFAOYNA-N
Synonym carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn
PubChem CID 23706213
SMILES [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC332611000
SDS View Documents
Thermo Scientific Chemicals
332611000
100 g Glass bottle
Each for $74.28
Only null left
 
AC332610010
SDS View Documents
Thermo Scientific Chemicals
332610010
1 kg Plastic bottle
Each for $218.43
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
SDS
Documents

Documents

RUO – Research Use Only