Learn More
Carbon disulfide, 99.9%, ACS reagent
CAS: 75-15- | CS2 | 76.13 g/mol
Supplier: Thermo Scientific Chemicals 447035000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Environmental benefits include:
- Recyclable
Specifications
Carbon disulfide | |
75-15- | |
46.0°C | |
−30°C | |
1.627 | |
1.262 | |
CS2 | |
151811 | |
76.13 | |
99.9% |
-111.0°C | |
1.2620g/mL | |
500 mL | |
Glass Bottle | |
99.9% min. (GC) | |
CS2 | |
0.363 mPa.s (20°C) | |
Solubility in water: 2.9g/L (20°C). Other solubilities: miscible with anhydrous methanol,ethanol,ether,,benzene,chloroform,carbon tetrachloride and oils | |
76.13 | |
ACS reagent |
Chemical Identifiers
75-15- | |
CS2 | |
76.13 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
Highly fla
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Danger
EINECSNumber : 200-843-6
RUO – Research Use Only