Please login to your online account to display your discounted pricing

Carbol-Fuchsin Solution, For Staining Acid-Fast Bacteria, LabChem™


Quantity 500mL

Manufacturer: lab chem  LC128801

 View more versions of this product

Catalog No. LC128801

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 94.11
Chemical Name or Material Carbol-Fuchsin Solution
CAS Max % 12.25
CAS 569-61-9
CAS Max % 4
CAS 67-63-0
CAS Max % 0.75
Identification Passes Test
Name Note For Staining Acid-Fast Bacteria
Physical Form Liquid
Packaging Amber Glass
Linear Formula C6H5O8
CAS 7732-18-5
Solubility Soluble in water
Synonym Kinyoun
Health Hazard 1 Danger
Decomposition Information Hydrogen chloride; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
Color Dark red
CAS Max % 63
Health Hazard 2 GHS H Statement
Harmful if swallowed or if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
May cause cancer.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve, liver, kidneys).
May cause damage to organs (liver, kidneys) through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.
CAS 64-17-5
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Use only outdoors or in a well-ventilated area.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Quantity 500mL
CAS Max % 19
Recommended Storage Room Temperature
CAS 108-95-2

Specifications

Chemical Name or Material Carbol-Fuchsin Solution
CAS 7732-18-5
CAS Max % 63
CAS 64-17-5
CAS Max % 19
CAS 108-95-2
CAS Max % 12.25
CAS 569-61-9
CAS Max % 4
CAS 67-63-0
CAS Max % 0.75
Name Note For Staining Acid-Fast Bacteria
Linear Formula C6H5O8
Formula Weight 94.11
Synonym Kinyoun
Accessories

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Use only outdoors or in a well-ventilated area.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed or if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
May cause cancer.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve, liver, kidneys).
May cause damage to organs (liver, kidneys) through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.

Room Temperature