Learn More
Cadmium nitrate tetrahydrate, 99+%
CAS: 10022-68-1 | CdH2N2O6 | 238.44 g/mol
$201.69 - $201.69
Chemical Identifiers
CAS | 10022-68-1 |
---|---|
Molecular Formula | CdH2N2O6 |
Molecular Weight (g/mol) | 238.44 |
MDL Number | MFCD00149626 |
InChI Key | IZIPRBSCGMISKX-UHFFFAOYSA-N |
Synonym | cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate |
PubChem CID | 56924536 |
ChEBI | CHEBI:86156 |
SMILES | [Cd++].O[N+]([O-])=O.O[N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC212342500
|
Thermo Scientific Chemicals
212342500 |
250 g | Glass bottle |
Each for $201.69
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10022-68-1 | |
238.44 | |
IZIPRBSCGMISKX-UHFFFAOYSA-N | |
56924536 | |
[Cd++].O[N+]([O-])=O.O[N+]([O-])=O |
CdH2N2O6 | |
MFCD00149626 | |
cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate | |
CHEBI:86156 |
Specifications
132°C | |
10022-68-1 | |
Crystals or Powder | |
99% min. (Complexometry) | |
Cd(NO3)2·4H2O | |
15, 1627 | |
(10% in water) Clear colorless | |
[Cd++].O[N+]([O-])=O.O[N+]([O-])=O | |
238.44 | |
CHEBI:86156 | |
≥99% | |
Cadmium nitrate tetrahydrate, p.a. |
59°C | |
Colorless or White | |
250 g | |
CdH2N2O6 | |
MFCD00149626 | |
cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate | |
IZIPRBSCGMISKX-UHFFFAOYSA-N | |
cadmium(2+) bis(nitric acid) | |
56924536 | |
308.46 | |
Glass bottle |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
Fatal if inhaled.
May cause cancer.
May cause genetic defects.
May damage fertility or the unborn child.
Causes damage to organs through prolonge
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/p
GHS Signal Word: Danger
RTECSNumber : EV1850000
RUO – Research Use Only