Learn More
Cadence Science⢠Laboratory Pipetting Needles
316 Stainless Steel Hub with 316 Stainless Steel Tubing
Supplier: Cadence Science⢠9870
Description
316 Stainless Steel Hub with 316 Stainless Steel Tubing
- Needles 14-825-16A through -16N and 14-818-106A through -106C all have standard nickel plated hubs and 304 stainless steel cannulae
- Needles 01-290-29 through -35 have 316 stainless steel Micro-Mate⢠Threaded Female Luer Hubs and 316 stainless steel cannulae
- All have 90° blunt ends
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
| Stainless Steel, Stainless Steel (Hub) | |
| 4 in. | |
| 12 Needles | |
| 90° Blunt | |
| Female Luer |
| 14 G | |
| Pipetting Needle | |
| 101.6 mm | |
| Manual and Automatic Volumetric Dispensers, Diluters, Pipetters |