missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientificâ„¢ C-Type Natriuretic Peptide (32-53), Human
For cellular and molecular biology applications
Specifications
Product Type | C-Type Natriuretic Peptide (32-53), human |
---|---|
Species | Human |
Molecular Weight (g/mol) | 2197.59 |
Content And Storage | -30°C to -10°C |
Sequence | Gly-Leu-Ser-Lys-Gly-Cys-Phe-Gly-Leu-Lys-Leu-Asp-Arg-Ile-Gly-Ser-Met-Ser-Gly-Leu-Gly-Cys (Disulfide bridge Cys6-Cys22) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ66540MCR
|
Thermo Scientificâ„¢
J66540MCR |
1 mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
C-Type Natriuretic Peptide (32-53), human | |
2197.59 | |
Gly-Leu-Ser-Lys-Gly-Cys-Phe-Gly-Leu-Lys-Leu-Asp-Arg-Ile-Gly-Ser-Met-Ser-Gly-Leu-Gly-Cys (Disulfide bridge Cys6-Cys22) | |
CNP-22 | |
HPLC |
Human | |
-30°C to -10°C | |
Solid | |
RUO - research use only |
RUO – Research Use Only