Learn More
Labcon North America U-Pette™ Bloiod Dispenser for Point of Care Test Cartridges, in Boxes
Supplier: Labcon North America 76305000009
Dispenser; Point of Care Test Cassettes; C-Pette™; Labcon; Blood collection tube dispenser for Point of Care testing instruments; Dispenses blood drops easy with no need to open collection tubes; Dispenses one drop of blood onto test cartridge or casssette; Fits standard glass or plastic blood tubes with rubber stoppers; Polystyrene USP Classs VI; Stainless steel canula; Not Autoclavable; 100 per box/5 boxes per case; Made in USA