missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green Indicator, 0.1% (w/v), Ricca Chemical

$78.66 - $169.81

Specifications

Chemical Name or Material Bromocresol Green Indicator
CAS 62625-32-5,7732-18-5
CAS Min % 98.0,0.1
CAS Max % 100.0,0.1
Concentration 0.1% (w/v)
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
1205-4
View Documents Promotion Details
Ricca Chemical Company
12054
120 mL Natural Poly Bottle
Each for $78.66
Only null left
 
1205-16
View Documents Promotion Details
Ricca Chemical Company
120516
500 mL Natural Poly Bottle
Each for $136.03
Only null left
 
R12050001A
View Documents Promotion Details
Ricca Chemical Company
R12050001A
1 L Natural Poly Bottle
Each for $169.81
Only null left
 
Description

Description

Specifications

Specifications

Bromocresol Green Indicator
98.0,0.1
0.1% (w/v)
Indicator
Aqueous Solution
>5.4
62625-32-5,7732-18-5
100.0,0.1
0.1% W/V
Natural Poly Bottle
Dark Blue
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'