missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocresol Green Indicator, 0.04% (w/v), Ricca Chemical
Supplier: Ricca Chemical Company RI12004
Specifications
Bromocresol Green Indicator | |
98.0,0.04 | |
0.04% (w/v) | |
Indicator | |
Aqueous Solution | |
>5.4 | |
Liquid |
62625-32-5,7732-18-5 | |
100.0,0.04 | |
0.04% W/V | |
Natural Poly Bottle | |
Dark Blue | |
120 mL |
Promotions
-
Ricca Chemical Company
Spend $100 or More, Get Up to $500 in Chemicals
Buy $100 or more of Ricca Chemical Company products, and get up to $500 more to spend on products at no additional cost.Expires: 06/30/26 Redemption Only