Learn More
BrandTech™ VACUUBRAND™ MD1C + AK + EK Oil-Free Chemistry Diaphragm Vacuum Pump
An excellent choice for rotary evaporation and other laboratory applications
Supplier:  BrandTech™ VACUUBRAND™ 20696620
Catalog No. 13-889-102
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
Suitable for benchtop rotary evaporators, and powerful enough for excellent results with high-boiling point solvents
  • Extremely low maintenance, no oil changes, no oil mist
  • Ultimate vacuum 1.5 Torr; free air capacity 25 liters per minute
  • Corrosion resistance
  • With an integrated gas ballast to provide consistent pump performance with condensed vapors
  • Adds solvent recovery, including an emissions condenser; compact three-stage design is suitable for benchtop rotary evaporation and gel drying with an SDS-PAGE concentration greater than 10%
Order Info
Included solvent recovery glassware for use in evaporative applications when no cold trap is used.
Specifications
Specifications
CEE plug
12.62 x 11.4 x 19 in. (32 x 29 x 48 cm)
MD1C+AK+EK
10.2 kg
60 Hz
48 cm
32 cm
29 cm
230 V
PTFE (Pump Head)
22.48 lb.
25 L/min.
19 in.
12.62 in.
11.4 in.
230 V
Product Suggestions
Product Suggestions