Learn More
Brady™ Steam Leak Tags
Durable Steam Leak Tags inform employees of the inspection history and status of equipment or materials.
Supplier:  Brady™ 86498
 View more versions of this product
Catalog No. 19-040-806
Only null left
Explore available promotions
Description
Description
  • The excellent write-on surface is ideal for adding on-the-spot customized notes and other information.
  • Polyester material and economical cardstock tags available.
  • Tags meet OSHA Standard 1910.145 for accident prevention.
  • Write-on polyester (B-837) laminate protects the graphics, wipes clean and withstands water, grease and extreme temperatures. Durable heavy duty polyester material with brass grommets exceeds OSHA's 50 lb pull force requirement for tagout devices.
  • Paper cardstock tags are recommended for indoor use only.
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Specifications
Specifications
ASTM D1593-09, ASTM D-638-08, OSHA 1910.145
Polyester
For lockout and tagout identification
3.9 Gardner
17.78cm
0.51 to 0.64 mm
-40°F to 175°F
10.16 cm
0.37 in.
Black, Red, Yellow
Header: Steam Leak, Front: Instructions/1. Fill Out Work Order/2. Enter Work Order Number On Back Side/3. Hang Tag On All Steam And Condensate Leaks, Back: Name/Date/Work Order Number
2 Years
7 in.
10 nylon fasteners
-40°C to 80°C
4 in.
7 x 4 in.
0.93 cm
Product Suggestions
Product Suggestions