Learn More
Brady™ Reversible Floor Signs
Reversible Floor Signs are an economical way to get your messages noticed.
Supplier:  Brady™ 92280
 View more versions of this product
Catalog No. 19-003-371
Only null left
Explore available promotions
Description
Description
  • Folds for easy storage when not in use
  • Each stand contains two different legends - just turn the stand inside out to display each one
Specifications
Specifications
ASTM D 1593, ASTM D 638, ASTM D 635, ASTM D 638
1 Ea.
Non-adhesive
20 in.
-40°F
12 in.
-40°C to 54°C
20 x 12 in.
Reversible Floor Stand
Polypropylene
1.5 lb./in.
Used for marking construction sites, temporary parking and asbestos abatement sites
130°F
0.16 in.
30.48 cm
-40°F to 130°F
Front Legend, Caution Slippery When Wet/ Back Legend, Caution Wet Floor
Black on Yellow
Product Suggestions
Product Suggestions