Learn More
Brady™ Keyed Alike Nylon Safety Padlocks with 1.5 in. Shackle Locks
Padlock offers unique keyway which breaks any current traveling form padlock shackle to cylinder
Supplier:  Brady™ 123333
 View more versions of this product
Catalog No. 19-103-452
Only null left
Explore available promotions
Description
Description
  •  Keyed-Alike padlock with non-conductive, spark resistant lock body
  • Compact and lightweight
  • Available in a variety of colors
Greener Choice Claims
  • Environmental benefits include:
    • Source Reduction
Specifications
Specifications
Red
Nylon
Padlock
Keyed Alike
3/Pk.
1.5 in.
Product Suggestions
Product Suggestions