missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Electrical Hazard Signs: WARNING - HIGH VOLTAGE KEEP AWAY
Identify electrical hazard and high voltage areas
$13.81 - $35.53
Specifications
Quantity | 1 Ea. |
---|---|
Mounting Type | Surface Mount |
Min. Temperature (English) | -40°F |
Displayed Text | Warning High Voltage Keep Away |
Product Type | Electrical Hazard Sign |
Catalog Number | Mfr. No. | Material | Height (English) | Width (English) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Height (English) | Width (English) | Price | Quantity | ||||
19-035-923
|
Brady™
22564 |
Plastic | 7 in. | 10 in. |
Each for $14.01
|
|
||||
19-035-926
|
Brady™
84944 |
Polyester | 10 in. | 14 in. |
Each for $35.53
|
|
||||
19-035-925
|
Brady™
84943 |
Polyester | 7 in. | 10 in. |
Each for $13.81
|
|
||||
Description
- Available in three materials - plastic, fiberglass and polyester
- Available in different widths and heights
- Black on orange
Specifications
1 Ea. | |
-40°F | |
Electrical Hazard Sign |
Surface Mount | |
Warning High Voltage Keep Away |