missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Electrical Hazard Signs: DANGER - HIGH VOLTAGE (Vertical)
Identify electrical hazard and high voltage areas
$19.19 - $19.19
Specifications
Material | Polyester |
---|---|
Quantity | 1 Ea. |
For Use With (Application) | Identification of fire alarms, extinguishers and emergency exits for both indoor/outdoor applications and in harsh environments where excellent abrasion and chemical resistance is required beyond a surface printed label |
Max. Temperature (English) | 230°F |
Min. Temperature (English) | -40°F |
Catalog Number | Mfr. No. | Height (English) | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Height (English) | Price | Quantity | ||||||
19-035893
|
Brady™
84878 |
10 in. |
N/A
|
|
||||||
19-035-894
|
Brady™
84879 |
14 in. |
N/A
|
|
||||||
Description
- Made up of self-sticking polyester
- Available in different widths and heights
- Black/red on white
Specifications
Polyester | |
Identification of fire alarms, extinguishers and emergency exits for both indoor/outdoor applications and in harsh environments where excellent abrasion and chemical resistance is required beyond a surface printed label | |
-40°F | |
-40°C to 110°C | |
DANGER High Voltage | |
Red on White |
1 Ea. | |
230°F | |
0.006 to 0.01 in. | |
-40°F to 230°F | |
Electrical Hazard Sign |