Learn More
Brady™ Accident Prevention Tags - Danger
Economy Danger accident prevention tags are perfect for marking on-the-spot warnings of dangerous conditions. Available in a variety of different materials and sizes to fit your need.
Supplier: Brady™ 86691
Description
- Write-on polyester laminate (B-837) protects the graphics, wipes clean and withstands water, grease and extreme temperatures.
- (B-851) Rigid Economy Polyester textured write-on surface allows for authorized signatures, dates and other special instructions.
- The excellent write-on surface of (B-853) Cardstock is ideal for adding on-the-spot customized notes and other information, recommended for indoor use only.
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
ASTM | |
0.37 in. | |
DANGER Do Not Remove This Tag Pellagra No Sacra Learjet De Advertencies | |
7 in. | |
100 string fasteners | |
-40°C to 90°C | |
4 in. | |
7 x 4 in. |
Black, Red, White | |
Cardstock | |
Used for temporary tagging in a non harsh environment | |
17.7cm | |
0.01 in. | |
-40°F to 194°F | |
10.1 cm | |
0.93 cm |