Learn More
MilliporeSigmaâ„¢ Supelcoâ„¢ Bottle-Type Reagent TLC Sprayer Set
Compressed-air sprayers for spraying TLC plates. Sets include a glass-sprayer head and a choice of either bottle or tube with hexfoot stand.
$219.09 - $309.09
Description
The extraction phase or adsorbent phase can be coated on to the solid phase as a thin film. Coating preparation is of three types; dipping, brush painting and spraying. Spraying method was widely used due to its stability and reusability.Specifications
TLC |