missing translation for 'onlineSavingsMsg'
Learn More

Accuform™ BLOCKADE Retractable Belt Tape Barrier Receiver Post

BLOCKADE retractable belt tape barrier receiver posts are for use at end of an extended belt tape barrier

Supplier:  Accuform™ PRB846BK

Catalog No. 19-119-214


Only null left
Explore available promotions

Description

Description

  • Post 2.5 in. diameter
  • 2 in. width woven polyester tape wound in a self-tension, spring-loaded unit
  • Tape extends up to 8 ft. - straight tension without sag or slack
  • Retracts smoothly back into head unit for storage
  • Safety mechanism in the spring-tension reduces whip action of belt braking
Specifications

Specifications

Black
Retractable Belt Tape Receiver Post
14 in.
38 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions