missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl chloroformate, 3M (50 wt%) solution in toluene, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 124491000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Coupling reagentGreener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Benzyl chloroformate | |
501-53-1,108-88-3,100-44-7 | |
53.0,55.0 | |
7°C | |
C8H7ClO2 | |
MFCD00000640 | |
06, 437 | |
1.01 | |
benzyl chloroformate, carbobenzoxy chloride, benzyl chlorocarbonate, cbz chloride, benzyloxycarbonyl chloride, carbobenzyloxy chloride, carbonochloridic acid, phenylmethyl ester, benzylcarbonyl chloride, cbz-cl, chloroformic acid, benzyl ester | |
HSDAJNMJOMSNEV-UHFFFAOYSA-N | |
benzyl carbonochloridate | |
10387 | |
45 to 53 wt% |
50 wt. % Solution in Toluene | |
45.0,47.0 | |
1.0100g/mL | |
Glass bottle | |
C6H5CH2OOCCl | |
100mL | |
01,109; 02,59; 15,22 | |
15, 1801 | |
Solubility in water: decomposes | |
C1=CC=C(C=C1)COC(=O)Cl | |
170.6 | |
170.6 |
RUO â Research Use Only