missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: SOB Medium
For cultivating recombinant strains of E. coli
Supplier: BD B244310
Specifications
<30°C | |
Super Optimal Broth Medium Medium | |
Tryptone 20g/L, Yeast extract 5g/L, Sodium Chloride 0.5g/L, Magnesium Sulfate 2.4g/L, Potassium Chloride 186mg/L | |
Bottle |
Dehydrated | |
Microbiology Media | |
Differentiation of enteric Bacilli on the basis of Sulfide production, indole formation and motility | |
500 g |