Learn More
BD Clay Adams™ Fixed-Angle Rotors for Dynac™ and Dynac III Centrifuges
For use with Dynac and Dynac III Centrifuges
Supplier: BD B420114
Description
- Six-, 12-, and 24-place fixed-angle rotors accept tubes from 3 to 15mL when equipped with buckets (sold separately) For optimum performance, the use of heavy-duty glassware is recommended in these rotors
- Environmental benefits include:
- Renewable Energy
Greener Choice Claims
Specifications
Fixed Angle Rotor | |
24 x 15 mL | |
Dynac™ and Dynac™ III Centrifuge |