missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Plated Media: Sabouraud Dextrose Agar (Deep Fill)
Used in qualitative procedures for cultivation of dermatophytes
Supplier: BD 221278
Description
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
- Deep-filled plates reduce effects of drying during prolonged incubation
Specifications
USP, ISO/DIS, 11014 | |
Solid | |
Sabouraud Dextrose Agar (deep fill) | |
Qualitative procedures for cultivation of Dermatophytes, Candida albicans | |
100/Pk. |
Light Beige | |
United States | |
Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 40g/L, Agar 15g/L | |
Plate |