missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Diphenylamine Sulfonate, 0.1% w/v for Chlorine, Reagents

Barium Diphenylamine Sulfonate, 0.1% w/v for Chlorine, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$113.40 - $256.43
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.02 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
IUPAC Name | water |
SMILES | O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
01-335-802
|
Labchem Inc
CB109900120A |
120 mL |
Each for $113.40
|
|
||||||
01-335-801
|
Labchem Inc
CB1099001A |
1 L |
Each for $256.43
|
|
||||||
01-335-800
|
Labchem Inc
CB109900500A |
500 mL |
Each for $191.57
|
|
||||||
Chemical Identifiers
7732-18-5 | |
18.02 | |
water |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
O |
Specifications
100°C | |
7732-18-5,1300-92-1 | |
120 mL | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
0.1% | |
Poly Bottle | |
Barium Diphenylamine Sulfonate, 0.1% w/v for Chlorine |
Colorless to Light Gray/Blue | |
Liquid | |
H2O | |
O | |
18.02 | |
Reagent | |
1.002 |