Learn More
MilliporeSigmaâ„¢ Supelcoâ„¢ Aura Mobile Phase Filter/Degasser Unit Reservoir
Easy-to-use Aura filter/degasser units simultaneously degas solvents and buffers and remove particles. Eliminates spurious peaks caused by gas bubbles in your detector, and prevent particle damage to check valves and other system components. A clean mobile phase also prolongs column life.
$1747.07 - $1747.07
Specifications
For Use With Solid Phase Extraction Disks (SPED)
View More Specs
Products 1
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
111003591
View Documents
MilliporeSigmaâ„¢ Supelcoâ„¢
58093
2 L
Each for $1,747.07
Only null left
Description
Description
Each unit includes a PTFE filtering/degassing assembly, ten 1.5 μm PTFE filter membranes, 40 in./1 m x 1/4 in. O.D. PTFE tubing, and a heavy wall, graduated, borosilicate glass solvent reservoir. Choose either a 1-liter or a 2-liter reservoir. The standard taper PTFE joint on the filtering assembly fits into Erlenmeyer flasks and other containers. The filtering assembly accepts any standard 47 mm filter membrane and is compatible with all solvents used in HPLC. Use with appropriate glassware and any vacuum pump.The Aura filter/degasser is particularly suitable for use with solid phase extraction disks (SPED). It ensures simple disc placement, large filtration area, small restriction to flow from the disk support element, and the convenience and advantages of the liquid inlet tube (allows sediments to be transferred last). Attaches to any vacuum line.
Specifications
Specifications
Solid Phase Extraction Disks (SPED)
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'