Learn More
Ammonium tetrachloroplatinate(II), 99.9% (metals basis), Pt 51% min
CAS: 13820-41-2 | Cl4H8N2Pt | 372.96 g/mol
Supplier: Thermo Scientific Chemicals 01104606
Description
Ammonium tetrachloroplatinate(II) is used in spectral analysis standard. It is also used in the preparation of platinum sponge and platinum catalytic agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Environmental benefits include:
- Recyclable
Specifications
Ammonium tetrachloroplatinate(II) | |
13820-41-2 | |
Crystalline | |
(metals basis) | |
(NH4)2PtCl4 | |
UN3288 | |
Soluble in water. | |
[NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Pt++] | |
372.96 | |
99.9% | |
Odorless |
145°C (decomposition) | |
Red | |
5 g | |
Cl4H8N2Pt | |
MFCD00010885 | |
ammonium tetrachloroplatinate ii, diammonium tetrachloroplatinate, ammonium platinous chloride, diammonium tetrachloroplatinate 2-, ammonium tetrachloroplatinate, bis ammonium tetrachloroplatinate 2-, ammonium chloroplatinate ii, unii-z5ny20a9s2, h8cl4n2pt, epitope id:151521 | |
QJIMNDWDOXTTBR-UHFFFAOYSA-L | |
platinum(2+) diammonium tetrachloride | |
372.98 | |
Pt 51% min. | |
2.936 g/mL |
Chemical Identifiers
13820-41-2 | |
372.96 | |
QJIMNDWDOXTTBR-UHFFFAOYSA-L | |
platinum(2+) diammonium tetrachloride |
Cl4H8N2Pt | |
MFCD00010885 | |
ammonium tetrachloroplatinate ii, diammonium tetrachloroplatinate, ammonium platinous chloride, diammonium tetrachloroplatinate 2-, ammonium tetrachloroplatinate, bis ammonium tetrachloroplatinate 2-, ammonium chloroplatinate ii, unii-z5ny20a9s2, h8cl4n2pt, epitope id:151521Show More | |
[NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Pt++] |
Safety and Handling
GHS H Statement
H301-H334-H318-H290-H315-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May be corrosive to metals.
Causes skin irritation.
May cause an allergic skin reaction.
P261-P264b-P270-P271-P272-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P330-P333+P313-P342+P311-P362-P501c
H301-H315-H317-H318-H334
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber : 237-499-1
RTECSNumber : TP1840000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only