missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate, Monobasic, Crystal, ACS, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor A1235500GM

 View more versions of this product

Catalog No. 18608767


Only null left
Explore available promotions

Description

Description

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications

Specifications

7722-76-1
3.8 to 4.4
H6NO4P
LFVGISIMTYGQHF-UHFFFAOYSA-N
phosphoric acid amine
98%
Poly Bottle
1
500 g
MFCD00003396
N.OP(O)(O)=O
115.03
ACS

Chemical Identifiers

7722-76-1
115.03
LFVGISIMTYGQHF-UHFFFAOYSA-N
N.OP(O)(O)=O
H6NO4P
MFCD00003396
phosphoric acid amine
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.