Learn More
Bioassay Systems ALP DETECTION REAGENT, 6 ML DFS Item
Supplier:  Bioassay Systems QFALP6ML
Detection Reagent. QuantiFluo ALP Reagent. Method: 360/450nm; Samples: Biological; Species: All; Procedure: 30 min; Size: 600 tests; Shelf Life: 2 years
Catalog No. 501078344
Only null left
Explore available promotions