Learn More
Allegro™ Bitrex Fit Test Kit: Solution
Offers qualitative OSHA compliant fit testing for disposable and reusable dust and mist respirators.
Supplier: Allegro™ 204112K
Description
Bitrex Sensitivity Solution and Bitrex Test Solution
- Box of 6 glass ampules each
- With pre-drilled plastic cap for easy pouring
- Plastic protection from broken glass
- One-time use
- Averages 3 to 5 fit tests per tube
- Contains 2.5cc of solution
Specifications
Test Solution | |
2041-12K | |
Test solution |
Allegro Bitrex Fit Test Kit (2041) | |
6/Pk. |