Learn More
MilliporeSigma™ Supelco™ Aldrich™ Flask-Type Sprayer
Connect using the barb end to a low-pressure gas or air source of <5 psi.
Supplier:  MilliporeSigma™ Supelco™ Z1291781EA
 View more versions of this product
Catalog No. 111010043
Only null left
Explore available promotions
Description
Description
By positioning your thumb over the vent hole, you can control the spray pattern.
Specifications
Specifications
250 mL
TLC
Aldrich
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC