missing translation for 'onlineSavingsMsg'
Learn More

AirClean™ Systems PowderSafe™ Type A Ductless Balance Enclosure

Based on SafeChange technology that minimizes threat of exposure to toxic powders during pre-filter replacement, the Airclean PowderSafe™ Type A Ductless Balance Enclosure is a smart addition to any lab environment.

Supplier:  AirClean™ Systems AC720C

 View more versions of this product

Catalog No. 36-100-4291


Only null left
Explore available promotions

Alert:

Requires an air filter in order to operate. Air filter must be purchased separately.


Description

Description

 

  • Provides effective and safe environment when using powder weighing equipment in conjunction with toxic compounds or chemicals
  • Electrostatically charged pre-filter is easily bagged out under negative pressure
  • AirSafe™ automatic safety controller
  • Controller allows operator to set desired face velocity thus minimizing airflow disturbance and maximizing operator protection
  • Portable-NO DUCTWORK REQUIRED
  • Constantly monitors airflow and filter conditions
  • Electrical cord access ports
  • Solid 3/8 in. polypropylene base
  • Tested with 3 and 4 place balances
  • Leak tested and certified before shipment
  • Available in 24 in., 32 in. and 48 in. widths
  • Quiet operation lessthan 49dB
  • 360° visibility
  • Compact ergonomic design takes up less space
  • Optional carbon filtration

 

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications

Specifications

Portable Type A Ductless Balance Enclosure
Polypropylene
24 x 48 x 30 in. (60.9 x 121.9 x 76.2 cm)
<49 dB
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions