Learn More
AirClean™ Systems PowderSafe Type B Enclosures
Choose the Airclean PowderSafe Type B Enclosure for powder weighing and containment technology.
$13608.89 - $17671.82
Catalog Number | Mfr. No. | Dimensions (D x W x H) | Width (English) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (D x W x H) | Width (English) | Price | Quantity | ||||
36-100-4292
|
AirClean™ Systems
AC730C |
53.3 x 78.7 x 45.7 cm (21 x 31 x 18 in.) | 32 in. |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
36-100-4293
|
AirClean™ Systems
AC740C |
55.8 x 119.3 x 45.7 cm (22 x 47 x 18 in.) | 48 in. |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- AirSafe™ automatic safety controller
- Solid polypropylene construction
- Effectively weigh to 5 decimal places
- Electrical cord access ports
- Continuous HEPA filter monitoring
- HEPASafe™ filter change technology provides a safe and effective means for replacement of both the pre-filter and primary HEPA filter
- Audible and visible alarms
- Real Time airflow display on LCD
- Portable
- Fluorescent light with one touch operation
- Modular electronics for easy service
- Optional sturdy mobile cart
Specifications
HEPA | |
33 in. | |
83.82 cm |