missing translation for 'onlineSavingsMsg'
Learn More

Activated charcoal, NORIT™ SA 2, decolorizing
GREENER_CHOICE

CAS: 7440-44-0 | C | 12 g/mol

$131.69 - $487.19

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC404035000
SDS View Documents
Thermo Scientific Chemicals
404035000
500 g Glass bottle
Each for $131.69
Only null left
 
AC404030025
SDS View Documents
Thermo Scientific Chemicals
404030025
2.5 kg Plastic drum
Each for $487.19
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Filter aid
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-153-3

RTECSNumber : FF5250100

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only