Learn More
Bioassay Systems EnzyChrom™ Aconitase Assay Kit. For quantitative determination of aconitase enzyme activity and high-throughput screening assays for aconitase modulator.
Supplier: Bioassay Systems EACO100
For quantitative determination of aconitase enzyme activity and high-throughput screening assays for aconitase modulator. Key Features: Fast and sensitive. Linear detection range (20 µL sample): 0.5 to 100 U/L for 20 min reaction. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handlingsystems for processing thousands of samples per day. Method: OD565nm. Samples: Biological samples (e.g. cell lysate, tissue homogenate, serum, etc.). Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 0.5 U/L.