missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, HPLC Grade, 99.7+% min, Thermo Scientific Chemicals

$234.78 - $234.78

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA22927K2
View Documents
Thermo Scientific Chemicals
022927K2
1 L
Each for $234.78
Only null left
 
Description

Description

Acetonitrile is used as a mobile phase in high performance liquid chromatography and liquid chromatography coupled with mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

H225-H302-H312-H332-H319: Highly flammable liquid and vapour. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes serious eye irritation.

DOTInformation : DOT Class: 3; Packing Group: II

EINECSNumber : 200-835-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only