missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, Gradient grade, 99.9% min, OmniSolv, MilliporeSigma™
Supplier: Technidata AX0142P1
Description
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Acetonitrile | |
Colorless | |
Glass Bottle (Pre-packed) | |
99.9% min. | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CC#N | |
41.053 | |
CHEBI:38472 |
75-05-8 | |
4 L | |
Liquid | |
C2H3N | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.05 |
Chemical Identifiers
75-05-8 | |
41.053 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecnShow More | |
CHEBI:38472 | |
CC#N |
C2H3N | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |