missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, 1% (v/v), Ricca Chemical

$58.13 - $78.75

Chemical Identifiers

CAS 7732-18-5
Molecular Formula C2H4O2
Molecular Weight (g/mol) Mixture
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
100-16
SDS View Documents Promotion Details
Ricca Chemical Company
RI10016
500 mL Natural Poly Bottle
Each for $58.13
Only null left
 
R10032
View Documents Promotion Details
Ricca Chemical Company
RI10032
1 L Natural Poly Bottle
Each for $78.75
Only null left
 
Description

Description

Specifications
Show More
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'