missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenone, 99+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 119200025

Catalog No. AC119200025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications

Specifications

9-Fluorenone
342.0°C
Authentic
Plastic bottle
2.5kg
9-fluorenone, 9h-fluoren-9-one, fluorenone, 9-oxofluorene, diphenylene ketone, unii-az9t83s2aq, ccris 593, 9h-fluorene-9-one, az9t83s2aq, 9-fluoreneone
YLQWCDOCJODRMT-UHFFFAOYSA-N
fluoren-9-one
10241
180.21
486-25-9
163°C
99% min. (GC)
C13H8O
07,465
Solubility in water: insoluble.
C1=CC=C2C(=C1)C3=CC=CC=C3C2=O
180.21
CHEBI:17922
99+%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 207-630-7

SDS Documents

Documents

One moment while we fetch your results.
Promotions

Promotions

RUO – Research Use Only