Learn More
6-Methyluracil, 97%

CAS: 626-48-2 | C5H6N2O2 | 126.12 g/mol
$48.49 - $708.70
Chemical Identifiers
CAS | 626-48-2 |
---|---|
Molecular Formula | C5H6N2O2 |
Molecular Weight (g/mol) | 126.12 |
MDL Number | MFCD00006028 |
InChI Key | SHVCSCWHWMSGTE-UHFFFAOYSA-N |
Synonym | 6-methyluracil, 2,4-dihydroxy-6-methylpyrimidine, pseudothymine, 6-methylpyrimidine-2,4 1h,3h-dione, 4-methyluracil, 6-methylpyrimidine-2,4-diol, 2,4 1h,3h-pyrimidinedione, 6-methyl, uracil, 6-methyl, 6-methyl-2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 6-methylShow More |
PubChem CID | 12283 |
ChEBI | CHEBI:74733 |
IUPAC Name | 6-methyl-1H-pyrimidine-2,4-dione |
SMILES | CC1=CC(=O)NC(=O)N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAB2419122
|
Thermo Scientific Chemicals
B2419122 |
100 g |
Each for $48.49
|
|
||||||
AAB2419136
|
Thermo Scientific Chemicals
B2419136 |
500 g |
Each for $158.03
|
|
||||||
AAB241910E
|
Thermo Scientific Chemicals
B241910E |
2500 g |
Each for $708.70
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
626-48-2 | |
126.12 | |
SHVCSCWHWMSGTE-UHFFFAOYSA-N | |
12283 | |
6-methyl-1H-pyrimidine-2,4-dione |
C5H6N2O2 | |
MFCD00006028 | |
6-methyluracil, 2,4-dihydroxy-6-methylpyrimidine, pseudothymine, 6-methylpyrimidine-2,4 1h,3h-dione, 4-methyluracil, 6-methylpyrimidine-2,4-diol, 2,4 1h,3h-pyrimidinedione, 6-methyl, uracil, 6-methyl, 6-methyl-2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 6-methylShow More | |
CHEBI:74733 | |
CC1=CC(=O)NC(=O)N1 |
Specifications
626-48-2 | |
C5H6N2O2 | |
100 g | |
14,6133 | |
SHVCSCWHWMSGTE-UHFFFAOYSA-N | |
6-methyl-1H-pyrimidine-2,4-dione | |
12283 | |
126.12 | |
6-Methyluracil |
317°C to 320°C | |
MFCD00006028 | |
115647 | |
6-methyluracil, 2,4-dihydroxy-6-methylpyrimidine, pseudothymine, 6-methylpyrimidine-2,4 1h,3h-dione, 4-methyluracil, 6-methylpyrimidine-2,4-diol, 2,4 1h,3h-pyrimidinedione, 6-methyl, uracil, 6-methyl, 6-methyl-2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 6-methyl | |
CC1=CC(=O)NC(=O)N1 | |
126.12 | |
CHEBI:74733 | |
97% |
Safety and Handling
GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
P201-P202-P281-P308+P313-P501c
H361f
EINECSNumber : 210-949-4
RTECSNumber : YR0700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only