missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5H-Chromeno[2,3-b]pyridin-5-one, 97%, Thermo Scientificâ„¢
$75.16 - $75.16
Chemical Identifiers
CAS | 6537-46-8 |
---|---|
Molecular Formula | C12H7NO2 |
MDL Number | MFCD00052253 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
BTB03140DA
|
Thermo Scientific Chemicals
BTB03140DA |
1 g |
Each for $75.16
|
|
||||||
Chemical Identifiers
6537-46-8 | |
C12H7NO2 | |
MFCD00052253 |
Specifications
6537-46-8 | |
100.0 | |
Amber glass bottle | |
MFCD00052253 | |
197.19 | |
5H-Chromeno[2, 3-b]pyridin-5-one |
96.5 | |
97% | |
C12H7NO2 | |
1 g | |
97% |