missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-thiophenecarboxaldehyde, 98%

CAS: 13679-70-4 | C6H6OS | 126.18 g/mol

$629.65 - $629.65

Chemical Identifiers

CAS 13679-70-4
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.18
MDL Number MFCD00005434
InChI Key VAUMDUIUEPIGHM-UHFFFAOYSA-N
Synonym 5-methyl-2-thiophenecarboxaldehyde, 5-methylthiophene-2-carboxaldehyde, 5-methyl-2-thiophenecarbaldehyde, 2-formyl-5-methylthiophene, 5-methyl-2-formylthiophene, 2-thiophenecarboxaldehyde, 5-methyl, 5-methylthiophene-2-aldehyde, 5-methyl-2-thenaldehyde, 5-methyl-2-thiophencarboxaldehyde, 5-methyl-2-thiophene carboxaldehydeShow MoreShow Less
PubChem CID 61663
IUPAC Name 5-methylthiophene-2-carbaldehyde
SMILES CC1=CC=C(S1)C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC127870500
SDS View Documents
Thermo Scientific Chemicals
127870500
50 mL Glass bottle
Each for $629.65
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 237-178-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only