missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorocytosine, 98+%

CAS: 2022-85-7 | C4H4FN3O | 129.09 g/mol

$77.15 - $77.15

Chemical Identifiers

CAS 2022-85-7
Molecular Formula C4H4FN3O
Molecular Weight (g/mol) 129.09
MDL Number MFCD00006035,MFCD00179326,MFCD03547958
InChI Key XRECTZIEBJDKEO-UHFFFAOYSA-N
Synonym 5-fluorocytosine, flucytosine, ancotil, ancobon, flucytosin, 5-fluorocystosine, fluocytosine, 5-fc, 5-fluorocytosin, fluorcytosine
PubChem CID 3366
ChEBI CHEBI:5100
SMILES NC1=C(F)C=NC(=O)N1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL16496MD
View Documents
Thermo Scientific Chemicals
L16496MD
250 mg
Each for $77.15
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
5-Fluorocytosine acts as an antidiabetic, antifungal and antimicrobial agent. It is useful for the treatment of serious infections arises due to susceptible strains of Candida or Cryptococcus neoformans and chromomycosis. Further, it is employed in studies on TMP biosynthesis.

Solubility
Soluble in water and hydrochloric acid.

Notes
Light Sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

2022-85-7
129.09
XRECTZIEBJDKEO-UHFFFAOYSA-N
3366
NC1=C(F)C=NC(=O)N1
C4H4FN3O
MFCD00006035,MFCD00179326,MFCD03547958
5-fluorocytosine, flucytosine, ancotil, ancobon, flucytosin, 5-fluorocystosine, fluocytosine, 5-fc, 5-fluorocytosin, fluorcytosine
CHEBI:5100

Specifications

2022-85-7
C4H4FN3O
250 mg
Light sensitive
5-fluorocytosine, flucytosine, ancotil, ancobon, flucytosin, 5-fluorocystosine, fluocytosine, 5-fc, 5-fluorocytosin, fluorcytosine
XRECTZIEBJDKEO-UHFFFAOYSA-N
6-amino-5-fluoro-1H-pyrimidin-2-one
3366
129.09
5-Fluorocytosine
∽297°C (decomposition)
MFCD00006035,MFCD00179326,MFCD03547958
127285
14,4125
Soluble in water and hydrochloric acid.
NC1=C(F)C=NC(=O)N1
129.09
CHEBI:5100
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341
Suspected of causing genetic defects.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 217-968-7

RTECSNumber : HA6040000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only